The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081559.2 | NP_001075028.1 | 5 - 6 | 642 | 87 |
NM_001318160.1 | NP_001305089.1 | 5 - 6 | 642 | 87 | |
NM_001318162.1 | NP_001305091.1 | 5 - 6 | 617 | 87 | |
NM_006693.3 | NP_006684.1 | 5 - 6 | 642 | 87 | |
XM_017011701.1 | XP_016867190.1 | 5 - 6 | 662 | 87 | |
GenBank mRNA | AK301745.1 | - | 5 - 6 | 572 | 87 |
BC003101.1 | - | 5 - 6 | 557 | 87 | |
BC050738.1 | - | 5 - 6 | 605 | 87 | |
BQ052009.1 | - | 5 - 6 | 604 | 87 | |
BX358595.2 | - | 5 - 6 | 616 | 86 | |
CR542161.1 | - | 5 - 6 | 499 | 87 | |
CX754488.1 | - | 5 - 6 | 610 | 87 | |
U79569.1 | - | 5 - 6 | 535 | 87 |