The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278192.1 | NP_001265121.1 | 19 - 20 | 2365 | 71 |
NM_001278193.1 | NP_001265122.1 | 18 - 19 | 2299 | 71 | |
NM_001278194.1 | NP_001265123.1 | 20 - 21 | 2532 | 71 | |
NM_198334.2 | NP_938148.1 | 21 - 22 | 2641 | 71 | |
NM_198335.3 | NP_938149.2 | 22 - 23 | 2707 | 71 | |
XM_017017412.1 | XP_016872901.1 | 19 - 20 | 2375 | 71 | |
XM_017017413.1 | XP_016872902.1 | 21 - 22 | 2757 | 71 | |
XM_017017414.1 | XP_016872903.1 | 21 - 22 | 2588 | 71 | |
GenBank mRNA | AF144074.1 | - | 22 - 23 | 2596 | 71 |
AK026548.1 | - | 10 - 11 | 1045 | 71 | |
AK026567.1 | - | 2 - 3 | 183 | 71 | |
AK026997.1 | - | 21 - 22 | 2532 | 71 | |
AK295806.1 | - | 18 - 19 | 2187 | 71 | |
AK299819.1 | - | 20 - 21 | 2415 | 71 | |
AK302752.1 | - | 19 - 20 | 2271 | 71 | |
AK314370.1 | - | 21 - 22 | 2530 | 71 | |
BC005405.1 | - | 4 - 5 | 621 | 71 | |
BC017433.2 | - | 11 - 12 | 1146 | 71 | |
BC017435.2 | - | 11 - 12 | 1146 | 71 | |
BC034439.1 | - | 12 - 13 | 1517 | 71 | |
BC065266.1 | - | 20 - 21 | 2408 | 71 | |
EU794613.1 | - | 21 - 22 | 2547 | 71 |