The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198334.2 | NP_938148.1 | 3 - 4 | 379 | 62 |
NM_198335.3 | NP_938149.2 | 3 - 4 | 379 | 62 | |
XM_017017412.1 | XP_016872901.1 | 1 - 2 | 113 | 62 | |
XM_017017413.1 | XP_016872902.1 | 3 - 4 | 442 | 62 | |
XM_017017414.1 | XP_016872903.1 | 3 - 4 | 273 | 62 | |
GenBank mRNA | AF144074.1 | - | 3 - 4 | 268 | 62 |
AJ000332.1 | - | 3 - 4 | 252 | 62 | |
AK026997.1 | - | 3 - 4 | 269 | 62 | |
AK314370.1 | - | 3 - 4 | 268 | 62 | |
BG720650.1 | - | 3 - 4 | 324 | 62 | |
D42041.1 | - | 3 - 4 | 250 | 62 | |
DC400833.1 | - | 3 - 4 | 379 | 62 | |
EU794613.1 | - | 3 - 4 | 285 | 62 |