The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005268208.1 | XP_005268265.1 | 16 - 17 | 2170 | 75 |
XM_005268209.3 | XP_005268266.1 | 16 - 17 | 1974 | 75 | |
XM_017021765.1 | XP_016877254.1 | 16 - 17 | 1937 | 75 | |
GenBank mRNA | AB082526.1 | - | 16 - 17 | 2106 | 75 |
AK128693.1 | - | 16 - 17 | 2574 | 75 | |
AL832633.2 | - | 10 - 11 | 1794 | 75 | |
AY048580.1 | - | 16 - 17 | 2000 | 75 | |
AY080896.1 | - | 16 - 17 | 2012 | 75 | |
BC009336.2 | - | 1 - 2 | 42 | 75 | |
BC093881.1 | - | 16 - 17 | 2081 | 75 | |
BC112101.1 | - | 16 - 17 | 2081 | 75 |