The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291931.1 | NP_001278860.1 | 9 - 10 | 883 | 60 |
NM_145003.4 | NP_659440.2 | 11 - 12 | 1540 | 60 | |
XM_011516921.1 | XP_011515223.1 | 10 - 11 | 1394 | 60 | |
XM_017013177.1 | XP_016868666.1 | 10 - 11 | 1397 | 60 | |
XM_017013178.1 | XP_016868667.1 | 10 - 11 | 1391 | 60 | |
XM_017013179.1 | XP_016868668.1 | 10 - 11 | 1397 | 60 | |
XM_017013180.1 | XP_016868669.1 | 10 - 11 | 1394 | 60 | |
GenBank mRNA | AK055726.1 | - | 11 - 12 | 1469 | 60 |
BC065226.1 | - | 9 - 10 | 802 | 60 | |
BC113556.1 | - | 10 - 11 | 1394 | 60 | |
BC126343.1 | - | 10 - 11 | 1397 | 60 | |
BC143684.1 | - | 10 - 11 | 1391 | 60 | |
BC143686.1 | - | 10 - 11 | 1397 | 60 |