Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSNARE1
Entrez Gene ID
203062
Gene Name
t-SNARE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 142212080 - 142403291 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145003.4 NP_659440.2 12 - 13 1628 130
XM_017013177.1 XP_016868666.1 11 - 12 1485 130
XM_017013178.1 XP_016868667.1 11 - 12 1479 130
GenBank mRNA AK055726.1 - 12 - 13 1557 130
BC113556.1 - 11 - 12 1482 130
BC126343.1 - 11 - 12 1485 130
BC143684.1 - 11 - 12 1479 130
BC143686.1 - 11 - 12 1485 130

View Full Product Details