Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TSNARE1
- Entrez Gene ID
-
203062
- Gene Name
- t-SNARE domain containing 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 142212080 - 142403291 on Build GRCh38
View Full Product Details