Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSNARE1
Entrez Gene ID
203062
Gene Name
t-SNARE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 142212080 - 142403291 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145003.4 NP_659440.2 7 - 8 1173 109
XM_011516912.1 XP_011515214.1 6 - 7 1027 109
XM_011516914.1 XP_011515216.1 7 - 8 1126 109
XM_011516915.2 XP_011515217.1 8 - 9 1212 109
XM_011516916.1 XP_011515218.1 7 - 8 1215 109
XM_011516917.2 XP_011515219.1 7 - 8 1052 109
XM_011516918.1 XP_011515220.1 5 - 6 877 109
XM_011516919.1 XP_011515221.1 6 - 7 1027 109
XM_011516920.1 XP_011515222.1 4 - 5 370 109
XM_011516922.1 XP_011515224.1 6 - 7 1027 109
XM_011516923.2 XP_011515225.1 2 - 3 1354 109
XM_017013175.1 XP_016868664.1 6 - 7 1027 109
XM_017013176.1 XP_016868665.1 6 - 7 1027 109
XM_017013177.1 XP_016868666.1 6 - 7 1027 109
XM_017013179.1 XP_016868668.1 6 - 7 1027 109
XM_017013180.1 XP_016868669.1 6 - 7 1027 109
XM_017013181.1 XP_016868670.1 6 - 7 1027 109
GenBank mRNA AK055726.1 - 7 - 8 1102 109
BC113556.1 - 6 - 7 1027 109
BC126343.1 - 6 - 7 1027 109
BC143686.1 - 6 - 7 1027 109

View Full Product Details