Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSNARE1
Entrez Gene ID
203062
Gene Name
t-SNARE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 142212080 - 142403291 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291931.1 NP_001278860.1 8 - 9 821 110
NM_145003.4 NP_659440.2 10 - 11 1478 110
XM_011516912.1 XP_011515214.1 9 - 10 1332 110
XM_011516913.2 XP_011515215.1 10 - 11 1520 110
XM_011516914.1 XP_011515216.1 10 - 11 1434 110
XM_011516915.2 XP_011515217.1 11 - 12 1520 110
XM_011516916.1 XP_011515218.1 10 - 11 1523 110
XM_011516917.2 XP_011515219.1 10 - 11 1360 110
XM_011516918.1 XP_011515220.1 8 - 9 1185 110
XM_011516919.1 XP_011515221.1 9 - 10 1335 110
XM_011516920.1 XP_011515222.1 7 - 8 678 110
XM_011516921.1 XP_011515223.1 9 - 10 1332 110
XM_011516922.1 XP_011515224.1 9 - 10 1335 110
XM_011516923.2 XP_011515225.1 5 - 6 1662 110
XM_017013175.1 XP_016868664.1 9 - 10 1335 110
XM_017013176.1 XP_016868665.1 9 - 10 1335 110
XM_017013177.1 XP_016868666.1 9 - 10 1335 110
XM_017013178.1 XP_016868667.1 9 - 10 1329 110
XM_017013179.1 XP_016868668.1 9 - 10 1335 110
XM_017013180.1 XP_016868669.1 9 - 10 1332 110
GenBank mRNA AK055726.1 - 10 - 11 1407 110
BC065226.1 - 8 - 9 740 110
BC113556.1 - 9 - 10 1332 110
BC126343.1 - 9 - 10 1335 110
BC143684.1 - 9 - 10 1329 110
BC143686.1 - 9 - 10 1335 110

View Full Product Details