The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022569.2 | NP_072091.1 | 4 - 5 | 1901 | 93 |
XM_017008545.1 | XP_016864034.1 | 3 - 4 | 899 | 93 | |
XM_017008546.1 | XP_016864035.1 | 2 - 3 | 205 | 93 | |
GenBank mRNA | AB036429.1 | - | 4 - 5 | 1901 | 93 |
AK289670.1 | - | 4 - 5 | 1879 | 93 | |
BC113075.2 | - | 2 - 3 | 490 | 93 |