Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK022678.1 - 1 1747 Chr. 10 - 124003054 - 124004974 [-] on Build GRCh38 88
GenBankmRNA AU149127.1 - 1 Chr. 10 - 124003054 - 124003646 [+] on Build GRCh38 88


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream carboxypeptidase X (M14 family), member 2 CPXM2 CPX2, UNQ676 -111.1kb Anti-sense Chr. - 123745636 - 123944094 on Build GRCh38
Downstream ornithine aminotransferase OAT GACR, HOGA, OATASE, OKT 393.7kb Anti-sense Chr. - 124397303 - 124418976 on Build GRCh38

View Full Product Details