Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO3
Entrez Gene ID
26273
Gene Name
F-box protein 3
Gene Aliases
FBA, FBX3
Chromosome Location
Chr.11: 33740937 - 33774531 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012175.3 NP_036307.2 8 - 9 961 82
NM_033406.2 NP_208385.1 8 - 9 961 82
XM_011519980.1 XP_011518282.1 8 - 9 967 82
XM_011519981.2 XP_011518283.1 8 - 9 967 82
XM_011519982.1 XP_011518284.1 9 - 10 1021 82
XM_017017533.1 XP_016873022.1 9 - 10 1021 82
GenBank mRNA AB209039.1 - 7 - 8 4346 82
AF176702.1 - 8 - 9 918 82
AK001943.1 - 8 - 9 952 82
AK128454.1 - 8 - 9 939 82
AK300721.1 - 3 - 4 499 82
AK304866.1 - 7 - 8 751 82
AK316286.1 - 3 - 4 267 82
AL136625.1 - 8 - 9 943 82
AL832826.1 - 7 - 8 1633 82
BC039291.1 - 8 - 9 2306 82
BC046110.1 - 8 - 9 947 82

View Full Product Details