The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286457.1 | NP_001273386.1 | 13 - 14 | 1495 | 104 |
NM_001286458.1 | NP_001273387.1 | 14 - 15 | 1670 | 104 | |
NM_001321858.1 | NP_001308787.1 | 13 - 14 | 1421 | 104 | |
NM_003470.2 | NP_003461.2 | 13 - 14 | 1626 | 104 | |
NR_135826.1 | 13 - 14 | 1449 | 104 | ||
XM_017023652.1 | XP_016879141.1 | 13 - 14 | 2054 | 104 | |
XM_017023653.1 | XP_016879142.1 | 13 - 14 | 1490 | 104 | |
GenBank mRNA | AK302481.1 | - | 14 - 15 | 1596 | 104 |
AK302771.1 | - | 13 - 14 | 1427 | 104 | |
AK302872.1 | - | 13 - 14 | 1449 | 104 | |
AK302912.1 | - | 14 - 15 | 1614 | 104 | |
AK316441.1 | - | 13 - 14 | 1496 | 104 | |
AY376241.1 | - | 13 - 14 | 1457 | 104 | |
Z72499.1 | - | 13 - 14 | 1626 | 104 |