The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002503.4 | NP_002494.2 | 1 - 2 | 281 | 72 |
GenBank mRNA | AB209784.1 | - | 1 - 2 | 247 | 72 |
AK290569.1 | - | 1 - 2 | 232 | 72 | |
BC015528.1 | - | 1 - 2 | 212 | 72 | |
BM008311.1 | - | 1 - 2 | 233 | 72 | |
BM449613.1 | - | 1 - 2 | 236 | 72 | |
BT006743.1 | - | 1 - 2 | 178 | 72 | |
DA633677.1 | - | 1 - 2 | 303 | 72 |