The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277225.1 | NP_001264154.1 | 2 - 3 | 287 | 101 |
NM_020973.4 | NP_066024.1 | 2 - 3 | 385 | 101 | |
NR_102355.1 | 2 - 3 | 385 | 101 | ||
NR_102357.1 | 2 - 3 | 287 | 101 | ||
GenBank mRNA | AB017913.1 | - | 2 - 3 | 334 | 101 |
AF317840.1 | - | 2 - 3 | 283 | 101 | |
AF323990.1 | - | 2 - 3 | 332 | 101 | |
AJ278964.1 | - | 2 - 3 | 283 | 101 | |
AK222963.1 | - | 2 - 3 | 332 | 101 | |
AK292746.1 | - | 2 - 3 | 332 | 101 | |
AK298377.1 | - | 2 - 3 | 287 | 101 | |
AK313189.1 | - | 2 - 3 | 337 | 101 | |
BC101829.1 | - | 2 - 3 | 320 | 101 | |
BC136264.1 | - | 2 - 3 | 320 | 101 |