The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014267.5 | NP_055082.1 | 4 - 5 | 497 | 64 |
XM_017017143.1 | XP_016872632.1 | 4 - 5 | 2756 | 64 | |
GenBank mRNA | AK057169.1 | - | 2 - 3 | 1565 | 64 |
AK093998.1 | - | 1 - 2 | 1310 | 64 | |
AK094549.1 | - | 1 - 2 | 2208 | 64 | |
AK315382.1 | - | 4 - 5 | 451 | 64 | |
BC007103.1 | - | 4 - 5 | 451 | 64 | |
BC016352.1 | - | 4 - 5 | 455 | 64 | |
BC020937.1 | - | 4 - 5 | 451 | 64 | |
BC103747.1 | - | 4 - 5 | 478 | 64 | |
BP210918.1 | - | 5 - 6 | 535 | 64 | |
BU661194.1 | - | 5 - 6 | 547 | 64 | |
BX648095.1 | - | 1 - 2 | 3781 | 64 | |
CR456882.1 | - | 4 - 5 | 319 | 64 | |
U51678.1 | - | 4 - 5 | 354 | 64 |