Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC25
Entrez Gene ID
548645
Gene Name
DnaJ heat shock protein family (Hsp40) member C25
Gene Aliases
bA16L21.2.1
Chromosome Location
Chr.9: 111631352 - 111654351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015882.2 NP_001015882.2 3 - 4 1012 136
NR_037148.1 4 - 5 1250 136
GenBank mRNA AF447177.1 - 4 - 5 1219 136
AK095452.1 - 4 - 5 1233 136
BC048318.1 - 3 - 4 983 136

View Full Product Details