Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PSRC1
- Entrez Gene ID
-
84722
- Gene Name
- proline and serine rich coiled-coil 1
- Gene Aliases
-
DDA3, FP3214
- Chromosome Location
-
Chr.1: 109279554 - 109283182 on Build GRCh38
View Full Product Details