The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198568.1 | NP_001185497.1 | 16 - 17 | 2469 | 63 |
NM_001198592.1 | NP_001185521.1 | 17 - 18 | 2168 | 63 | |
NM_139247.3 | NP_640340.2 | 17 - 18 | 2164 | 63 | |
GenBank mRNA | AF497516.1 | - | 16 - 17 | 2056 | 63 |
AK056745.1 | - | 5 - 6 | 430 | 63 | |
AK122714.1 | - | 17 - 18 | 2164 | 63 | |
AK126468.1 | - | 15 - 16 | 1890 | 63 | |
BC117473.1 | - | 16 - 17 | 2145 | 63 | |
BC117475.1 | - | 16 - 17 | 2145 | 63 | |
BC143947.1 | - | 16 - 17 | 2177 | 63 | |
BX248285.1 | - | 17 - 18 | 2135 | 63 | |
BX537489.1 | - | 10 - 11 | 3380 | 63 |