The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198568.1 | NP_001185497.1 | 20 - 21 | 3009 | 71 |
NM_001198592.1 | NP_001185521.1 | 21 - 22 | 2708 | 71 | |
NM_139247.3 | NP_640340.2 | 21 - 22 | 2704 | 71 | |
GenBank mRNA | AF497516.1 | - | 20 - 21 | 2596 | 71 |
AK056745.1 | - | 9 - 10 | 970 | 71 | |
AK122714.1 | - | 21 - 22 | 2704 | 71 | |
AK126468.1 | - | 16 - 17 | 3279 | 71 | |
BC117473.1 | - | 20 - 21 | 2685 | 71 | |
BC117475.1 | - | 20 - 21 | 2685 | 71 | |
BC143947.1 | - | 20 - 21 | 2717 | 71 | |
BX248285.1 | - | 21 - 22 | 2675 | 71 |