The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198568.1 | NP_001185497.1 | 21 - 22 | 3158 | 91 |
NM_001198592.1 | NP_001185521.1 | 22 - 23 | 2857 | 91 | |
NM_139247.3 | NP_640340.2 | 22 - 23 | 2853 | 91 | |
GenBank mRNA | AF497516.1 | - | 21 - 22 | 2745 | 91 |
AK056745.1 | - | 10 - 11 | 1119 | 91 | |
AK122714.1 | - | 22 - 23 | 2853 | 91 | |
AK126468.1 | - | 17 - 18 | 3428 | 91 | |
AK131333.1 | - | 12 - 13 | 2464 | 91 | |
BC117473.1 | - | 21 - 22 | 2834 | 91 | |
BC117475.1 | - | 21 - 22 | 2834 | 91 | |
BC143947.1 | - | 21 - 22 | 2866 | 91 | |
BX248285.1 | - | 22 - 23 | 2824 | 91 | |
BX537489.1 | - | 15 - 16 | 4729 | 91 |