The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198568.1 | NP_001185497.1 | 24 - 25 | 3501 | 63 |
NM_001198592.1 | NP_001185521.1 | 25 - 26 | 3200 | 63 | |
NM_139247.3 | NP_640340.2 | 25 - 26 | 3196 | 63 | |
GenBank mRNA | AF088070.1 | - | 2 - 3 | 175 | 63 |
AF497516.1 | - | 24 - 25 | 3088 | 63 | |
AK056745.1 | - | 13 - 14 | 1462 | 63 | |
AK122714.1 | - | 25 - 26 | 3196 | 63 | |
AK126468.1 | - | 20 - 21 | 3771 | 63 | |
AK131333.1 | - | 15 - 16 | 2807 | 63 | |
BC015982.2 | - | 2 - 3 | 243 | 63 | |
BC117473.1 | - | 24 - 25 | 3177 | 63 | |
BC117475.1 | - | 24 - 25 | 3177 | 63 | |
BC143947.1 | - | 24 - 25 | 3209 | 63 | |
BX248285.1 | - | 25 - 26 | 3167 | 63 | |
BX537489.1 | - | 18 - 19 | 5072 | 63 |