The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005257.5 | NP_005248.2 | 1 - 2 | 247 | 87 |
GenBank mRNA | BC027891.1 | - | 1 - 2 | 229 | 87 |
CN280495.1 | - | 1 - 2 | 233 | 87 | |
HY052912.1 | - | 1 - 2 | 247 | 87 | |
X95701.1 | - | 1 - 2 | 241 | 87 |