The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282948.1 | NP_001269877.1 | 3 - 4 | 783 | 110 |
NM_001318153.1 | NP_001305082.1 | 3 - 4 | 783 | 110 | |
NM_001318154.1 | NP_001305083.1 | 4 - 5 | 725 | 110 | |
NM_001322252.1 | NP_001309181.1 | 3 - 4 | 780 | 110 | |
NM_032776.2 | NP_116165.1 | 4 - 5 | 894 | 110 | |
XM_011539503.1 | XP_011537805.1 | 3 - 4 | 387 | 110 | |
XM_011539508.2 | XP_011537810.1 | 3 - 4 | 387 | 110 | |
XM_017015897.1 | XP_016871386.1 | 3 - 4 | 382 | 110 | |
XM_017015898.1 | XP_016871387.1 | 4 - 5 | 575 | 110 | |
XM_017015899.1 | XP_016871388.1 | 4 - 5 | 471 | 110 | |
XM_017015900.1 | XP_016871389.1 | 4 - 5 | 875 | 110 | |
GenBank mRNA | AK027280.1 | - | 4 - 5 | 739 | 110 |
BC143722.1 | - | 3 - 4 | 626 | 110 | |
EF068222.1 | - | 6 - 7 | 851 | 110 |