The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282948.1 | NP_001269877.1 | 4 - 5 | 904 | 137 |
NM_001318153.1 | NP_001305082.1 | 4 - 5 | 904 | 137 | |
NM_001318154.1 | NP_001305083.1 | 5 - 6 | 846 | 137 | |
NM_001322252.1 | NP_001309181.1 | 4 - 5 | 901 | 137 | |
NM_001322254.1 | NP_001309183.1 | 3 - 4 | 798 | 137 | |
NM_001322258.1 | NP_001309187.1 | 4 - 5 | 740 | 137 | |
NM_032776.2 | NP_116165.1 | 5 - 6 | 1015 | 137 | |
NR_134512.1 | 2 - 3 | 684 | 137 | ||
XM_011539503.1 | XP_011537805.1 | 4 - 5 | 508 | 137 | |
XM_011539508.2 | XP_011537810.1 | 4 - 5 | 508 | 137 | |
XM_017015897.1 | XP_016871386.1 | 4 - 5 | 503 | 137 | |
XM_017015898.1 | XP_016871387.1 | 5 - 6 | 696 | 137 | |
XM_017015899.1 | XP_016871388.1 | 5 - 6 | 592 | 137 | |
XM_017015900.1 | XP_016871389.1 | 5 - 6 | 996 | 137 | |
XM_017015901.1 | XP_016871390.1 | 4 - 5 | 539 | 137 | |
XM_017015902.1 | XP_016871391.1 | 4 - 5 | 800 | 137 | |
XM_017015903.1 | XP_016871392.1 | 3 - 4 | 348 | 137 | |
GenBank mRNA | AK027280.1 | - | 5 - 6 | 860 | 137 |
BC143722.1 | - | 4 - 5 | 747 | 137 | |
BX537954.1 | - | 2 - 3 | 649 | 137 | |
EF068222.1 | - | 7 - 8 | 972 | 137 |