The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021942.5 | NP_068761.4 | 4 - 5 | 650 | 62 |
NM_199053.2 | NP_951008.1 | 4 - 5 | 650 | 62 | |
XM_017008537.1 | XP_016864026.1 | 4 - 5 | 650 | 62 | |
XM_017008538.1 | XP_016864027.1 | 4 - 5 | 650 | 62 | |
XM_017008539.1 | XP_016864028.1 | 3 - 4 | 395 | 62 | |
GenBank mRNA | AK096345.1 | - | 4 - 5 | 624 | 62 |
AL833451.1 | - | 3 - 4 | 458 | 62 | |
AL833571.1 | - | 4 - 5 | 629 | 62 | |
BC051724.1 | - | 4 - 5 | 594 | 62 | |
BX538044.1 | - | 4 - 5 | 604 | 62 | |
BX647127.1 | - | 4 - 5 | 621 | 62 | |
BX648499.1 | - | 4 - 5 | 625 | 62 |