Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC42BPG
Entrez Gene ID
55561
Gene Name
CDC42 binding protein kinase gamma
Gene Aliases
DMPK2, HSMDPKIN, KAPPA-200, MRCKG, MRCKgamma
Chromosome Location
Chr.11: 64823809 - 64844685 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017525.2 NP_059995.2 16 - 17 1954 96
XM_011545155.2 XP_011543457.1 16 - 17 2069 96
XM_011545156.2 XP_011543458.1 16 - 17 2049 96
XM_011545157.2 XP_011543459.1 16 - 17 2070 96
XM_011545158.2 XP_011543460.1 16 - 17 2070 96
XM_011545159.2 XP_011543461.1 16 - 17 2070 96
XM_011545160.2 XP_011543462.1 17 - 18 2021 96
XM_011545161.2 XP_011543463.1 16 - 17 2070 96
XM_017017996.1 XP_016873485.1 16 - 17 2069 96
XM_017017997.1 XP_016873486.1 16 - 17 2070 96
XM_017017998.1 XP_016873487.1 16 - 17 2070 96
GenBank mRNA AY648038.1 - 16 - 17 1954 96

View Full Product Details