The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243474.1 | NP_001230403.1 | 7 - 8 | 2145 | 70 |
NM_001243476.2 | NP_001230405.1 | 11 - 12 | 2555 | 70 | |
NM_052851.2 | NP_443083.1 | 7 - 8 | 2145 | 70 | |
NM_178006.3 | NP_821074.1 | 7 - 8 | 2208 | 70 | |
NM_178007.2 | NP_821075.1 | 7 - 8 | 2115 | 70 | |
XM_011535299.2 | XP_011533601.1 | 10 - 11 | 2301 | 70 | |
XM_017020834.1 | XP_016876323.1 | 7 - 8 | 2313 | 70 | |
XM_017020835.1 | XP_016876324.1 | 9 - 10 | 2234 | 70 | |
GenBank mRNA | AK091804.1 | - | 7 - 8 | 2117 | 70 |
AK094704.1 | - | 6 - 7 | 1106 | 70 | |
AK095114.1 | - | 3 - 4 | 828 | 70 | |
AK312888.1 | - | 7 - 8 | 2201 | 70 | |
AL049801.1 | - | 7 - 8 | 2101 | 70 | |
AY082589.1 | - | 7 - 8 | 2203 | 70 | |
AY082590.1 | - | 7 - 8 | 2127 | 70 | |
AY082591.1 | - | 7 - 8 | 2101 | 70 | |
AY366448.1 | - | 7 - 8 | 2143 | 70 | |
BX647695.1 | - | 11 - 12 | 2515 | 70 |