Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHL8
Entrez Gene ID
57563
Gene Name
kelch like family member 8
Gene Aliases
-
Chromosome Location
Chr.4: 87160103 - 87220608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292003.1 NP_001278932.1 7 - 8 1754 90
NM_001292006.1 NP_001278935.1 7 - 8 1483 90
NM_001292007.1 NP_001278936.1 6 - 7 1162 90
NM_020803.4 NP_065854.3 7 - 8 1711 90
NR_120447.1 5 - 6 975 90
GenBank mRNA AB037799.2 - 6 - 7 1394 90
AK128827.1 - 7 - 8 1754 90
BC041384.2 - 7 - 8 1661 90
BC041901.1 - 5 - 6 831 90
BX538173.1 - 7 - 8 1628 90
BX640727.1 - 7 - 8 1621 90
BX640744.1 - 7 - 8 1412 90
BX647454.1 - 7 - 8 1646 90
BX648367.1 - 5 - 6 679 90
CR749265.1 - 6 - 7 1080 90

View Full Product Details