The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002190.2 | NP_002181.1 | 1 - 2 | 76 | 88 |
GenBank mRNA | BC066252.1 | - | 1 - 2 | 74 | 88 |
BC066253.1 | - | 1 - 2 | 74 | 88 | |
BC067503.1 | - | 1 - 2 | 74 | 88 | |
BC067504.1 | - | 1 - 2 | 74 | 88 | |
BC067505.1 | - | 1 - 2 | 74 | 88 | |
U32659.1 | - | 1 - 2 | 84 | 88 | |
Z58820.1 | - | 1 - 2 | 73 | 88 |