The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032268.4 | NP_115644.1 | 2 - 3 | 1182 | 57 |
XM_017023793.1 | XP_016879282.1 | 2 - 3 | 1156 | 57 | |
GenBank mRNA | AF378524.1 | - | 2 - 3 | 854 | 57 |
AK294434.1 | - | 2 - 3 | 854 | 57 | |
AL136903.1 | - | 2 - 3 | 847 | 57 | |
BC007235.1 | - | 2 - 3 | 854 | 57 |