The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031415.2 | NP_113603.1 | 3 - 4 | 1285 | 74 |
XM_011517158.1 | XP_011515460.1 | 3 - 4 | 1025 | 74 | |
XM_011517159.1 | XP_011515461.1 | 3 - 4 | 1025 | 74 | |
XM_011517161.1 | XP_011515463.1 | 3 - 4 | 1025 | 74 | |
XM_011517162.1 | XP_011515464.1 | 3 - 4 | 1025 | 74 | |
XM_011517163.2 | XP_011515465.1 | 2 - 3 | 252 | 74 | |
XM_011517167.1 | XP_011515469.1 | 3 - 4 | 1025 | 74 | |
XM_011517168.1 | XP_011515470.1 | 3 - 4 | 1025 | 74 | |
XM_017013644.1 | XP_016869133.1 | 3 - 4 | 1025 | 74 | |
XM_017013645.1 | XP_016869134.1 | 1 - 2 | 164 | 74 | |
GenBank mRNA | AB042405.1 | - | 3 - 4 | 1294 | 74 |
BC035321.1 | - | 3 - 4 | 830 | 74 | |
BC063595.1 | - | 3 - 4 | 813 | 74 |