Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSDMC
Entrez Gene ID
56169
Gene Name
gasdermin C
Gene Aliases
MLZE
Chromosome Location
Chr.8: 129705903 - 129786888 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031415.2 NP_113603.1 6 - 7 1606 102
XM_011517158.1 XP_011515460.1 6 - 7 1346 102
XM_011517159.1 XP_011515461.1 6 - 7 1346 102
XM_011517161.1 XP_011515463.1 6 - 7 1346 102
XM_011517162.1 XP_011515464.1 6 - 7 1346 102
XM_011517163.2 XP_011515465.1 5 - 6 573 102
XM_017013644.1 XP_016869133.1 6 - 7 1346 102
XM_017013645.1 XP_016869134.1 4 - 5 379 102
GenBank mRNA AB042405.1 - 6 - 7 1615 102
BC035321.1 - 6 - 7 1151 102
BC063595.1 - 6 - 7 1134 102

View Full Product Details