Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF16
Entrez Gene ID
7564
Gene Name
zinc finger protein 16
Gene Aliases
HZF1, KOX9
Chromosome Location
Chr.8: 144930358 - 144950888 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029976.2 NP_001025147.2 3 - 4 382 140
NM_006958.2 NP_008889.2 2 - 3 296 140
XM_005272341.2 XP_005272398.1 2 - 3 273 140
XM_011517298.1 XP_011515600.1 2 - 3 398 140
GenBank mRNA AK127625.1 - 2 - 3 296 140
AK226016.1 - 2 - 3 266 140
AL834451.1 - 1 - 2 213 140
BC010996.2 - 2 - 3 230 140
DQ117529.1 - 3 - 4 382 140

View Full Product Details