Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFSD5
Entrez Gene ID
84975
Gene Name
major facilitator superfamily domain containing 5
Gene Aliases
hsMOT2
Chromosome Location
Chr.12: 53251251 - 53254406 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032889.4 NP_116278.3 1 - 2 182 144
GenBank mRNA AK074684.1 - 1 - 2 179 144
DA428788.1 - 1 - 2 182 144
DA695979.1 - 1 - 2 181 146

View Full Product Details