The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099735.1 | NP_001093205.1 | 4 - 5 | 532 | 59 |
NM_001099736.1 | NP_001093206.1 | 4 - 5 | 536 | 59 | |
NM_001825.2 | NP_001816.2 | 5 - 6 | 696 | 59 | |
GenBank mRNA | AK122769.1 | - | 4 - 5 | 496 | 59 |
AK313293.1 | - | 4 - 5 | 523 | 59 | |
BC029140.1 | - | 4 - 5 | 513 | 59 | |
CR450315.1 | - | 3 - 4 | 454 | 59 | |
DC308656.1 | - | 4 - 5 | 536 | 59 | |
J05401.1 | - | 4 - 5 | 652 | 59 |