The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099735.1 | NP_001093205.1 | 7 - 8 | 66 | |
NM_001099736.1 | NP_001093206.1 | 7 - 8 | 66 | ||
NM_001825.2 | NP_001816.2 | 8 - 9 | 66 | ||
GenBank mRNA | AK122769.1 | - | 7 - 8 | 66 | |
AK313293.1 | - | 7 - 8 | 66 | ||
AW300490.1 | - | 3 - 4 | 523 | 66 | |
BC029140.1 | - | 7 - 8 | 66 | ||
CR450315.1 | - | 6 - 7 | 66 | ||
J05401.1 | - | 7 - 8 | 66 |