The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025207.4 | NP_079483.3 | 6 - 7 | 2032 | 100 |
NM_201398.2 | NP_958800.1 | 7 - 8 | 1588 | 100 | |
GenBank mRNA | AF481877.1 | - | 6 - 7 | 1340 | 100 |
AF520568.1 | - | 6 - 7 | 1953 | 100 | |
AK025979.1 | - | 7 - 8 | 1516 | 100 | |
BC020253.1 | - | 5 - 6 | 867 | 100 | |
BC021096.1 | - | 7 - 8 | 1506 | 100 | |
BC032323.1 | - | 7 - 8 | 1523 | 100 | |
DQ458779.1 | - | 6 - 7 | 1631 | 100 | |
U79241.1 | - | 5 - 6 | 1183 | 100 |