Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C21orf59
Entrez Gene ID
56683
Gene Name
chromosome 21 open reading frame 59
Gene Aliases
C21orf48, CILD26, FBB18, Kur
Chromosome Location
Chr.21: 32601674 - 32612608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021254.2 NP_067077.1 3 - 4 742 127
GenBank mRNA AF282851.1 - 3 - 4 490 127
AK055328.1 - 4 - 5 679 127
AK097566.1 - - 1643 127
AK098577.1 - 3 - 4 742 127
AK223318.1 - 3 - 4 720 127
AK225697.1 - 3 - 4 720 127
BC000709.2 - 3 - 4 495 127
FJ649644.1 - 3 - 4 1670 127

View Full Product Details