The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282403.1 | NP_001269332.1 | 5 - 6 | 653 | 89 |
NM_001282404.1 | NP_001269333.1 | 5 - 6 | 610 | 89 | |
NM_005918.3 | NP_005909.2 | 6 - 7 | 779 | 89 | |
GenBank mRNA | AF047470.1 | - | 6 - 7 | 723 | 89 |
AK095803.1 | - | 5 - 6 | 544 | 89 | |
AK290779.1 | - | 6 - 7 | 713 | 89 | |
AK293460.1 | - | 5 - 6 | 511 | 89 | |
AK316587.1 | - | 6 - 7 | 662 | 89 | |
BC001917.1 | - | 6 - 7 | 665 | 89 | |
CR536548.1 | - | 6 - 7 | 637 | 89 | |
DQ402957.1 | - | 5 - 6 | 574 | 89 |