The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252598.1 | NP_001239527.1 | 1 - 2 | 137 | 89 |
GenBank mRNA | AY780649.1 | - | 1 - 2 | 73 | 89 |
BP321569.1 | - | 1 - 2 | 114 | 89 |