The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145666.1 | NP_001139138.1 | 20 - 21 | 2949 | 106 |
NM_001145667.1 | NP_001139139.1 | 21 - 22 | 2982 | 106 | |
NM_012201.5 | NP_036333.2 | 21 - 22 | 2982 | 106 | |
NR_027264.1 | 22 - 23 | 3091 | 106 | ||
NR_027265.1 | 16 - 17 | 1979 | 106 | ||
GenBank mRNA | AK025457.1 | - | 18 - 19 | 2375 | 106 |
AK026972.1 | - | 12 - 13 | 1341 | 106 | |
AK027032.1 | - | 22 - 23 | 3052 | 106 | |
AK131501.1 | - | 3 - 4 | 1777 | 106 | |
AK172806.1 | - | 4 - 5 | 443 | 106 | |
AK304156.1 | - | 20 - 21 | 2919 | 106 | |
BC031836.1 | - | 16 - 17 | 1933 | 106 | |
BC060822.1 | - | 21 - 22 | 2953 | 106 | |
U28811.1 | - | 21 - 22 | 2952 | 106 | |
U64791.1 | - | 21 - 22 | 2935 | 106 |