The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145666.1 | NP_001139138.1 | 4 - 5 | 1000 | 112 |
NM_001145667.1 | NP_001139139.1 | 5 - 6 | 1033 | 112 | |
NM_012201.5 | NP_036333.2 | 5 - 6 | 1033 | 112 | |
NR_027264.1 | 5 - 6 | 1033 | 112 | ||
GenBank mRNA | AK025457.1 | - | 2 - 3 | 426 | 112 |
AK027032.1 | - | 5 - 6 | 994 | 112 | |
AK304156.1 | - | 4 - 5 | 970 | 112 | |
BC060822.1 | - | 5 - 6 | 1004 | 112 | |
U28811.1 | - | 5 - 6 | 1006 | 112 | |
U64791.1 | - | 5 - 6 | 986 | 112 |