Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EZH1
Entrez Gene ID
2145
Gene Name
enhancer of zeste 1 polycomb repressive complex 2 subunit
Gene Aliases
KMT6B
Chromosome Location
Chr.17: 42700275 - 42745096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321079.1 NP_001308008.1 13 - 14 1626 102
NM_001321081.1 NP_001308010.1 14 - 15 1690 102
NM_001321082.1 NP_001308011.1 13 - 14 1597 102
NM_001991.4 NP_001982.2 14 - 15 1717 102
XM_005257145.2 XP_005257202.1 12 - 13 1472 102
XM_011524517.2 XP_011522819.1 13 - 14 1565 102
XM_017024350.1 XP_016879839.1 13 - 14 1544 102
XM_017024351.1 XP_016879840.1 12 - 13 1451 102
XM_017024352.1 XP_016879841.1 11 - 12 1302 102
GenBank mRNA AB002386.1 - 13 - 14 1629 102
AB004818.1 - 13 - 14 1635 102
AK295626.1 - 13 - 14 1579 102
AK295853.1 - 12 - 13 1417 102
AK299887.1 - 13 - 14 1532 102
AK304835.1 - 13 - 14 1530 102
AK309121.1 - 13 - 14 1537 102
AK310399.1 - 12 - 13 1409 102
AK310556.1 - 6 - 7 761 102
AL833159.1 - 14 - 15 1637 102
BC015882.1 - 14 - 15 1651 102
BT009782.1 - 12 - 13 1529 102
L38934.1 - 7 - 8 733 102
U50315.1 - 13 - 14 1635 102

View Full Product Details