Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EZH1
Entrez Gene ID
2145
Gene Name
enhancer of zeste 1 polycomb repressive complex 2 subunit
Gene Aliases
KMT6B
Chromosome Location
Chr.17: 42700275 - 42745096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321079.1 NP_001308008.1 2 - 3 209 120
NM_001321081.1 NP_001308010.1 3 - 4 300 120
NM_001321082.1 NP_001308011.1 3 - 4 300 120
NM_001991.4 NP_001982.2 3 - 4 300 120
XM_005257145.2 XP_005257202.1 2 - 3 175 120
XM_011524517.2 XP_011522819.1 2 - 3 175 120
XM_017024352.1 XP_016879841.1 3 - 4 244 120
GenBank mRNA AB002386.1 - 2 - 3 212 120
AB004818.1 - 2 - 3 218 120
AK295626.1 - 2 - 3 162 120
AK295853.1 - 3 - 4 241 120
AK299887.1 - 3 - 4 235 120
AK309121.1 - 3 - 4 241 120
AK311237.1 - 3 - 4 244 120
AL833159.1 - 3 - 4 247 120
BC015882.1 - 3 - 4 234 120
BT009782.1 - 1 - 2 112 120
DC333017.1 - 3 - 4 257 120
U50315.1 - 2 - 3 218 120

View Full Product Details