The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321079.1 | NP_001308008.1 | 3 - 4 | 342 | 63 |
NM_001991.4 | NP_001982.2 | 4 - 5 | 433 | 63 | |
XM_017024352.1 | XP_016879841.1 | 4 - 5 | 377 | 63 | |
GenBank mRNA | AB002386.1 | - | 3 - 4 | 345 | 63 |
AB004818.1 | - | 3 - 4 | 351 | 63 | |
AK295626.1 | - | 3 - 4 | 295 | 63 | |
AK304835.1 | - | 3 - 4 | 246 | 63 | |
AK309121.1 | - | 4 - 5 | 374 | 63 | |
BC015882.1 | - | 4 - 5 | 367 | 63 | |
BT009782.1 | - | 2 - 3 | 245 | 63 | |
DC333017.1 | - | 4 - 5 | 390 | 63 | |
DC423942.1 | - | 4 - 5 | 433 | 63 | |
U50315.1 | - | 3 - 4 | 351 | 63 |