The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321079.1 | NP_001308008.1 | 8 - 9 | 1021 | 104 |
NM_001321081.1 | NP_001308010.1 | 9 - 10 | 1085 | 104 | |
NM_001321082.1 | NP_001308011.1 | 8 - 9 | 992 | 104 | |
NM_001991.4 | NP_001982.2 | 9 - 10 | 1112 | 104 | |
XM_005257145.2 | XP_005257202.1 | 7 - 8 | 867 | 104 | |
XM_011524517.2 | XP_011522819.1 | 8 - 9 | 960 | 104 | |
XM_017024350.1 | XP_016879839.1 | 8 - 9 | 939 | 104 | |
XM_017024351.1 | XP_016879840.1 | 7 - 8 | 846 | 104 | |
GenBank mRNA | AB002386.1 | - | 8 - 9 | 1024 | 104 |
AB004818.1 | - | 8 - 9 | 1030 | 104 | |
AK295626.1 | - | 8 - 9 | 974 | 104 | |
AK295853.1 | - | 7 - 8 | 812 | 104 | |
AK299887.1 | - | 8 - 9 | 927 | 104 | |
AK304835.1 | - | 8 - 9 | 925 | 104 | |
AK309121.1 | - | 8 - 9 | 932 | 104 | |
AK310399.1 | - | 7 - 8 | 804 | 104 | |
AK311237.1 | - | 8 - 9 | 909 | 104 | |
AL833159.1 | - | 9 - 10 | 1032 | 104 | |
BC015882.1 | - | 9 - 10 | 1046 | 104 | |
BT009782.1 | - | 7 - 8 | 924 | 104 | |
U50315.1 | - | 8 - 9 | 1030 | 104 |