The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023012.5 | NP_075388.2 | 6 - 7 | 888 | 73 |
NR_036434.1 | 7 - 8 | 966 | 73 | ||
NR_036435.1 | 7 - 8 | 988 | 73 | ||
NR_036436.1 | 6 - 7 | 910 | 73 | ||
XM_005253601.1 | XP_005253658.1 | 6 - 7 | 888 | 73 | |
XM_005253602.2 | XP_005253659.1 | 6 - 7 | 859 | 73 | |
XM_005253604.1 | XP_005253661.1 | 7 - 8 | 970 | 73 | |
XM_011538688.1 | XP_011536990.1 | 6 - 7 | 888 | 73 | |
XM_011538689.1 | XP_011536991.1 | 9 - 10 | 1143 | 73 | |
XM_017019830.1 | XP_016875319.1 | 6 - 7 | 888 | 73 | |
XM_017019831.1 | XP_016875320.1 | 7 - 8 | 965 | 73 | |
XM_017019832.1 | XP_016875321.1 | 9 - 10 | 1121 | 73 | |
XM_017019833.1 | XP_016875322.1 | 7 - 8 | 915 | 73 | |
GenBank mRNA | AB212664.1 | - | 6 - 7 | 874 | 73 |
AF161432.1 | - | 2 - 3 | 363 | 73 | |
AK001883.1 | - | 6 - 7 | 845 | 73 | |
AK023985.1 | - | 3 - 4 | 478 | 73 | |
AK056114.1 | - | 7 - 8 | 911 | 73 | |
AK056256.1 | - | 6 - 7 | 845 | 73 | |
BC008684.1 | - | 6 - 7 | 798 | 73 | |
BC067773.1 | - | 6 - 7 | 858 | 73 | |
BX640711.1 | - | 6 - 7 | 834 | 73 | |
BX647884.1 | - | 6 - 7 | 877 | 73 |