The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261412.1 | NP_001248341.1 | 6 - 7 | 637 | 64 |
NM_001261413.1 | NP_001248342.1 | 5 - 6 | 631 | 64 | |
NM_006400.4 | NP_006391.1 | 5 - 6 | 646 | 64 | |
XM_011537761.2 | XP_011536063.1 | 6 - 7 | 598 | 64 | |
GenBank mRNA | AK055682.1 | - | 5 - 6 | 509 | 64 |
AK096242.1 | - | 7 - 8 | 505 | 64 | |
AK098794.1 | - | 6 - 7 | 491 | 64 | |
AK222693.1 | - | 5 - 6 | 509 | 64 | |
AK292364.1 | - | 7 - 8 | 524 | 64 | |
AK314705.1 | - | 5 - 6 | 488 | 64 | |
AY189155.1 | - | 6 - 7 | 369 | 64 | |
BC000718.2 | - | 5 - 6 | 495 | 64 | |
BC009468.1 | - | 5 - 6 | 421 | 64 | |
BC014083.2 | - | 5 - 6 | 469 | 64 | |
FJ224315.1 | - | 5 - 6 | 456 | 64 | |
U50733.1 | - | 5 - 6 | 456 | 64 |