Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR27
Entrez Gene ID
253769
Gene Name
WD repeat domain 27
Gene Aliases
-
Chromosome Location
Chr.6: 169426420 - 169702071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK293148.1 - 7 - 8 1218 92
BX648854.1 - 4 - 5 416 92

View Full Product Details